Product Details

SNP ID
rs185474285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4914976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCCCTGGAGGCGATGGACAGCA[C/T]GGTGCGCAGGATCAGAGCATAAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51A7 PubMed Links

Gene Details

Gene
OR51A7
Gene Name
olfactory receptor family 51 subfamily A member 7
There are no transcripts associated with this gene.

Gene
OR51G1
Gene Name
olfactory receptor family 51 subfamily G member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR51G2
Gene Name
olfactory receptor family 51 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005238.1 688 Missense Mutation ATG,GTG M230V NP_001005238.1

View Full Product Details