Product Details

SNP ID
rs185557554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103707365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCAGTCCACGTAGAGTTTCCG[A/C]GTGGTTTCTCCTTCCCTTAGAAGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLRA4 PubMed Links

Gene Details

Gene
GLRA4
Gene Name
glycine receptor alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024452.2 1654 Silent Mutation ACG,ACT T411T NP_001019623.2
NM_001172285.1 1654 Intron NP_001165756.1
Gene
TMEM31
Gene Name
transmembrane protein 31
There are no transcripts associated with this gene.

View Full Product Details