Product Details
- SNP ID
-
rs185587002
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89686734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATGGCGGAGCCAGATCTGGAGTG[C/T]GAGCAGATCCGTCTGAAGTGTATTC
- Phenotype
-
MIM: 603464
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDK10
PubMed Links
Gene Details
- Gene
- CDK10
- Gene Name
- cyclin dependent kinase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098533.2 |
71 |
UTR 5 |
|
|
NP_001092003.2 |
NM_001160367.1 |
71 |
UTR 5 |
|
|
NP_001153839.1 |
NM_052987.3 |
71 |
UTR 5 |
|
|
NP_443713.2 |
NM_052988.4 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
NP_443714.3 |
XM_006721308.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_006721371.1 |
XM_006721310.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_006721373.1 |
XM_011523405.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_011521707.1 |
XM_011523406.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_011521708.1 |
XM_011523407.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_011521709.1 |
XM_011523408.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_011521710.1 |
XM_011523410.2 |
71 |
UTR 5 |
|
|
XP_011521712.1 |
XM_011523411.2 |
71 |
UTR 5 |
|
|
XP_011521713.1 |
XM_011523412.1 |
71 |
UTR 5 |
|
|
XP_011521714.1 |
XM_011523413.2 |
71 |
Intron |
|
|
XP_011521715.1 |
XM_011523414.2 |
71 |
Intron |
|
|
XP_011521716.1 |
XM_011523415.2 |
71 |
UTR 5 |
|
|
XP_011521717.1 |
XM_011523416.2 |
71 |
UTR 5 |
|
|
XP_011521718.1 |
XM_011523417.2 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_011521719.1 |
XM_017023806.1 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_016879295.1 |
XM_017023807.1 |
71 |
Silent Mutation |
TGC,TGT |
C8C |
XP_016879296.1 |
XM_017023808.1 |
71 |
UTR 5 |
|
|
XP_016879297.1 |
XM_017023809.1 |
71 |
UTR 5 |
|
|
XP_016879298.1 |
XM_017023810.1 |
71 |
Intron |
|
|
XP_016879299.1 |
- Gene
- SPATA2L
- Gene Name
- spermatogenesis associated 2 like
There are no transcripts associated with this gene.
View Full Product Details