Product Details

SNP ID
rs185491243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56189864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCCCGGTGTGTTTTCAGGTGA[C/T]GCTTGAATGTCCCCAGCTGACGGAA
Phenotype
MIM: 611178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
There are no transcripts associated with this gene.

Gene
ZSCAN5B
Gene Name
zinc finger and SCAN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080456.2 1631 Missense Mutation CAT,CGT H484R NP_001073925.2
XM_005258864.3 1631 Missense Mutation CAT,CGT H537R XP_005258921.2
XM_006723189.3 1631 Missense Mutation CAT,CGT H484R XP_006723252.1

View Full Product Details