Product Details

SNP ID
rs185217735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46408176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAGCAATGTGACAAGTATGAC[A/G]CCCAGGCACTCTCAGCCCAGCTGGT
Phenotype
MIM: 608379
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCRL2 PubMed Links

Gene Details

Gene
CCRL2
Gene Name
C-C motif chemokine receptor like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130910.1 382 Missense Mutation ACC,GCC T45A NP_001124382.1
NM_003965.4 382 Missense Mutation ACC,GCC T33A NP_003956.2
XM_011534208.1 382 Missense Mutation ACC,GCC T33A XP_011532510.1
XM_011534209.1 382 Missense Mutation ACC,GCC T33A XP_011532511.1
XM_017007436.1 382 Missense Mutation ACC,GCC T33A XP_016862925.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details