Product Details

SNP ID
rs185398866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42161697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTCACCTGTTCCCAATCTTC[A/T]TCTTACACACCATGCACACCTTCTC
Phenotype
MIM: 612188
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 2715 Missense Mutation AAG,ATG K857M NP_001288067.1
NM_015289.3 2715 Missense Mutation AAG,ATG K846M NP_056104.2
XM_011521403.2 2715 Missense Mutation AAG,ATG K857M XP_011519705.1
XM_011521404.2 2715 Missense Mutation AAG,ATG K846M XP_011519706.1
XM_017022036.1 2715 Missense Mutation AAG,ATG K857M XP_016877525.1
XM_017022037.1 2715 Missense Mutation AAG,ATG K846M XP_016877526.1
XM_017022038.1 2715 Missense Mutation AAG,ATG K722M XP_016877527.1
XM_017022039.1 2715 Missense Mutation AAG,ATG K489M XP_016877528.1

View Full Product Details