Product Details
- SNP ID
-
rs186027780
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143376631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCTGGAGGAGCTCAGCGGTGGC[A/G]TGGACCCTGGCCGGCATGGGAGGTG
- Phenotype
-
MIM: 601031
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RHPN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3812439] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RHPN1
- Gene Name
- rhophilin Rho GTPase binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052924.2 |
934 |
Missense Mutation |
ATG,GTG |
M95V |
NP_443156.2 |
XM_005250772.3 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_005250829.1 |
XM_005250773.3 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_005250830.1 |
XM_006716502.3 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_006716565.1 |
XM_011516817.2 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_011515119.1 |
XM_011516818.1 |
934 |
Missense Mutation |
ATG,GTG |
M131V |
XP_011515120.1 |
XM_011516819.2 |
934 |
Missense Mutation |
ATG,GTG |
M95V |
XP_011515121.1 |
XM_011516820.2 |
934 |
Missense Mutation |
ATG,GTG |
M83V |
XP_011515122.1 |
XM_011516821.2 |
934 |
Missense Mutation |
ATG,GTG |
M46V |
XP_011515123.1 |
XM_011516822.2 |
934 |
Missense Mutation |
ATG,GTG |
M46V |
XP_011515124.1 |
XM_011516823.2 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_011515125.1 |
XM_017013013.1 |
934 |
Intron |
|
|
XP_016868502.1 |
XM_017013014.1 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_016868503.1 |
XM_017013015.1 |
934 |
Missense Mutation |
ATG,GTG |
M46V |
XP_016868504.1 |
XM_017013016.1 |
934 |
Missense Mutation |
ATG,GTG |
M148V |
XP_016868505.1 |
- Gene
- RHPN1-AS1
- Gene Name
- RHPN1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details