Product Details

SNP ID
rs186133014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126632584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGCCACGATTTTCATTAAAGC[A/G]AGGGATGATCCCAAGACGTTATGTC
Phenotype
MIM: 604924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHAX PubMed Links

Gene Details

Gene
PHAX
Gene Name
phosphorylated adaptor for RNA export
There are no transcripts associated with this gene.

Gene
TEX43
Gene Name
testis expressed 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207408.1 138 Missense Mutation CAA,CGA Q42R NP_997291.1

View Full Product Details