Product Details

SNP ID
rs186096500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25849841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAAGACATTGATATAAATCCCT[A/G]ACTGACACAATGTGCTTAATCCGCA
Phenotype
MIM: 611034
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC17A3 PubMed Links

Gene Details

Gene
SLC17A3
Gene Name
solute carrier family 17 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098486.1 1111 Missense Mutation TCA,TTA S412L NP_001091956.1
NM_006632.3 1111 Missense Mutation TCA,TTA S334L NP_006623.2

View Full Product Details