Product Details

SNP ID
rs185922526
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:44810270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGTATTTGCCCATCGGTGCTT[C/G]TTGGCCTGTAGATGCAACTTCTTCC
Phenotype
MIM: 602913 MIM: 603673 MIM: 611713
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 483 Missense Mutation TTC,TTG F48L NP_001130009.1
XM_006710386.3 483 Missense Mutation TTC,TTG F48L XP_006710449.1
XM_011540803.2 483 Missense Mutation TTC,TTG F48L XP_011539105.1
XM_011540805.2 483 Missense Mutation TTC,TTG F48L XP_011539107.1
XM_017000447.1 483 Missense Mutation TTC,TTG F48L XP_016855936.1
XM_017000448.1 483 Missense Mutation TTC,TTG F48L XP_016855937.1
XM_017000449.1 483 Missense Mutation TTC,TTG F48L XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
There are no transcripts associated with this gene.

Gene
PTCH2
Gene Name
patched 2
There are no transcripts associated with this gene.

Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 483 Intron NP_001013654.1

View Full Product Details