Product Details

SNP ID
rs185690068
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18237435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGCTGCTATCTGCTTTCTTAA[C/T]ATTGGCACTTTTCACAGATTTTTTA
Phenotype
MIM: 125264
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DEK PubMed Links

Gene Details

Gene
DEK
Gene Name
DEK proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134709.1 935 Missense Mutation ATT,GTT I248V NP_001128181.1
NM_003472.3 935 Missense Mutation ATT,GTT I282V NP_003463.1
XM_017011273.1 935 Missense Mutation ATT,GTT I282V XP_016866762.1

View Full Product Details