Product Details

SNP ID
rs185654375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52065300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTTGGTGTTTAGTGAGGCAAGAC[C/G]GCCGCCCAAACGCCTTGCCACATTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF841 PubMed Links

Gene Details

Gene
ZNF841
Gene Name
zinc finger protein 841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136499.1 4637 Missense Mutation CCG,CGG P861R NP_001129971.1
NM_001321349.1 4637 Missense Mutation CCG,CGG P745R NP_001308278.1
XM_011526833.2 4637 Intron XP_011525135.1
XM_017026652.1 4637 Intron XP_016882141.1
XM_017026653.1 4637 Missense Mutation CCG,CGG P862R XP_016882142.1
XM_017026654.1 4637 Missense Mutation CCG,CGG P862R XP_016882143.1
XM_017026655.1 4637 Intron XP_016882144.1
XM_017026656.1 4637 Missense Mutation CCG,CGG P861R XP_016882145.1
XM_017026657.1 4637 Intron XP_016882146.1
XM_017026658.1 4637 Intron XP_016882147.1
XM_017026659.1 4637 Intron XP_016882148.1
XM_017026660.1 4637 Intron XP_016882149.1
XM_017026661.1 4637 Intron XP_016882150.1
XM_017026662.1 4637 Intron XP_016882151.1
XM_017026663.1 4637 Intron XP_016882152.1

View Full Product Details