Product Details

SNP ID
rs186248794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120606162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGCCTTTTTATCCAACCCTAA[A/G]ATTACTTCACAAATGTAAGGTTTTT
Phenotype
MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCTS1 PubMed Links

Gene Details

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 408 Missense Mutation AAA,AGA K84R NP_001131026.1
NM_014060.2 408 Missense Mutation AAA,AGA K83R NP_054779.1

View Full Product Details