Product Details
- SNP ID
-
rs186564713
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:93575090 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCTGCAGCATCTCTGTAGGCTC[A/T]CCATTAACAAATGTACCGGTGCCAT
- Phenotype
-
MIM: 605117
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SOCS2
PubMed Links
Gene Details
- Gene
- SOCS2
- Gene Name
- suppressor of cytokine signaling 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270467.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_001257396.1 |
NM_001270468.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_001257397.1 |
NM_001270469.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_001257398.1 |
NM_001270470.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_001257399.1 |
NM_001270471.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_001257400.1 |
NM_003877.4 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
NP_003868.1 |
XM_011538929.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_011537231.1 |
XM_011538935.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_011537237.1 |
XM_011538936.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_011537238.1 |
XM_017020147.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875636.1 |
XM_017020148.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875637.1 |
XM_017020149.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875638.1 |
XM_017020150.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875639.1 |
XM_017020151.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875640.1 |
XM_017020152.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875641.1 |
XM_017020153.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875642.1 |
XM_017020154.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875643.1 |
XM_017020155.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875644.1 |
XM_017020156.1 |
996 |
Missense Mutation |
ACC,TCC |
T170S |
XP_016875645.1 |
- Gene
- SOCS2-AS1
- Gene Name
- SOCS2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details