Product Details
- SNP ID
-
rs186312303
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:157092839 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCTGAGGGACAAGAGCCAAAGC[C/T]TCTCACCCGCACCCTGTCCCAGCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ETV3L
PubMed Links
Gene Details
- Gene
- ETV3L
- Gene Name
- ETS variant 3 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004341.2 |
1181 |
Missense Mutation |
AAG,AGG |
K299R |
NP_001004341.1 |
View Full Product Details