Product Details

SNP ID
rs186353882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:88722661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGACTGAGGGACTTTCCCTTTC[A/G]TCCGGCGAAGGTCTGGTGAGTCCAA
Phenotype
MIM: 600662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEF2C PubMed Links

Gene Details

Gene
MEF2C
Gene Name
myocyte enhancer factor 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131005.2 1480 Silent Mutation NP_001124477.1
NM_001193347.1 1480 Silent Mutation NP_001180276.1
NM_001193348.1 1480 Silent Mutation NP_001180277.1
NM_001193349.1 1480 Silent Mutation NP_001180278.1
NM_001193350.1 1480 Silent Mutation NP_001180279.1
NM_001308002.1 1480 Silent Mutation NP_001294931.1
NM_002397.4 1480 Silent Mutation NP_002388.2
XM_005248511.2 1480 Silent Mutation XP_005248568.1
XM_006714619.2 1480 Silent Mutation XP_006714682.1
XM_006714625.3 1480 Silent Mutation XP_006714688.1
XM_011543396.2 1480 Silent Mutation XP_011541698.1
XM_011543397.2 1480 Silent Mutation XP_011541699.1
XM_011543400.1 1480 Silent Mutation XP_011541702.1
XM_011543401.1 1480 Silent Mutation XP_011541703.1
XM_017009475.1 1480 Silent Mutation XP_016864964.1
XM_017009476.1 1480 Silent Mutation XP_016864965.1
XM_017009477.1 1480 Silent Mutation XP_016864966.1
XM_017009478.1 1480 Silent Mutation XP_016864967.1
XM_017009479.1 1480 Silent Mutation XP_016864968.1
XM_017009480.1 1480 Silent Mutation XP_016864969.1
XM_017009481.1 1480 Silent Mutation XP_016864970.1
XM_017009482.1 1480 Silent Mutation XP_016864971.1
XM_017009483.1 1480 Silent Mutation XP_016864972.1

View Full Product Details