Product Details

SNP ID
rs186390684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104491026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGGGAACTTGATCGTCCTAATG[C/T]AAAACGTACCCAGCGTAGCCCCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP12 PubMed Links

Gene Details

Gene
LRP12
Gene Name
LDL receptor related protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135703.2 2354 Missense Mutation ACA,GCA T724A NP_001129175.1
NM_013437.4 2354 Missense Mutation ACA,GCA T743A NP_038465.1
Gene
MIR548A3
Gene Name
microRNA 548a-3
There are no transcripts associated with this gene.

View Full Product Details