Product Details

SNP ID
rs187224046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53483064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGGGCGCCCCCAACTGCATCCC[C/G]TGTAAAGGTAGGACTCCTTCTTCCC
Phenotype
MIM: 136470
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FST PubMed Links

Gene Details

Gene
FST
Gene Name
follistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006350.3 438 Silent Mutation CCC,CCG P90P NP_006341.1
NM_013409.2 438 Silent Mutation CCC,CCG P90P NP_037541.1
XM_005248400.4 438 Silent Mutation CCC,CCG P90P XP_005248457.1
XM_005248401.4 438 Silent Mutation CCC,CCG P90P XP_005248458.1
XM_005248402.4 438 Silent Mutation CCC,CCG P90P XP_005248459.1
XM_005248403.4 438 Silent Mutation CCC,CCG P90P XP_005248460.1
XM_011543099.2 438 Silent Mutation CCC,CCG P85P XP_011541401.1
XM_017008954.1 438 Silent Mutation CCC,CCG P90P XP_016864443.1
XM_017008955.1 438 Silent Mutation CCC,CCG P12P XP_016864444.1

View Full Product Details