Product Details
- SNP ID
-
rs187205661
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:152109147 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTCGCGGAATTTTCTGTCACGC[C/T]CTTGGCGGCGCAGCGGCTGTTCCTC
- Phenotype
-
MIM: 190370
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCHH
PubMed Links
Gene Details
- Gene
- TCHH
- Gene Name
- trichohyalin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007113.3 |
4165 |
Missense Mutation |
GAG,GGG |
E1357G |
NP_009044.2 |
View Full Product Details