Product Details

SNP ID
rs186995901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56064324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCAGTCCTGATGGCCCGGCAT[G/T]GGTTACCGCTGCTGCCCCTGCTGTC
Phenotype
MIM: 104613 MIM: 607940
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT6A PubMed Links

Gene Details

Gene
CCT6A
Gene Name
chaperonin containing TCP1 subunit 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009186.1 101 Intron NP_001009186.1
NM_001762.3 101 Intron NP_001753.1
Gene
SNORA15
Gene Name
small nucleolar RNA, H/ACA box 15
There are no transcripts associated with this gene.

Gene
SUMF2
Gene Name
sulfatase modifying factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042469.1 101 Missense Mutation GGG,TGG G24W NP_001035934.2
NM_001042470.1 101 Missense Mutation GGG,TGG G24W NP_001035935.2
NM_001130069.2 101 Missense Mutation GGG,TGG G24W NP_001123541.1
NM_001146333.1 101 UTR 5 NP_001139805.1
NM_015411.2 101 Missense Mutation GGG,TGG G24W NP_056226.2
XM_011515254.2 101 Missense Mutation GGG,TGG G24W XP_011513556.1
XM_017011939.1 101 Missense Mutation GGG,TGG G24W XP_016867428.1
XM_017011940.1 101 Missense Mutation GGG,TGG G24W XP_016867429.1

View Full Product Details