Product Details

SNP ID
rs187661233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58334968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACGCCTGCGCTTCCGGCACTTC[C/T]GCTATGAGGAGGCATCTGGTCCACA
Phenotype
MIM: 165260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6806 PubMed Links

Gene Details

Gene
MIR6806
Gene Name
microRNA 6806
There are no transcripts associated with this gene.

Gene
ZSCAN22
Gene Name
zinc finger and SCAN domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321116.1 313 Missense Mutation CGC,TGC R56C NP_001308045.1
NM_001321117.1 313 Missense Mutation CGC,TGC R56C NP_001308046.1
NM_181846.2 313 Missense Mutation CGC,TGC R56C NP_862829.1
XM_006723192.3 313 Missense Mutation CGC,TGC R56C XP_006723255.1
XM_011526917.2 313 Missense Mutation CGC,TGC R56C XP_011525219.1
XM_011526918.2 313 Intron XP_011525220.1
XM_017026746.1 313 Missense Mutation CGC,TGC R56C XP_016882235.1
XM_017026747.1 313 UTR 5 XP_016882236.1

View Full Product Details