Product Details

SNP ID
rs187620078
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72074337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAATGGCTATGTGGAGCACTTG[G/T]TTCGCTACCAGTGTAAGAACTACTA
Phenotype
MIM: 140210
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HPR PubMed Links
Additional Information
For this assay, SNP(s) [rs7202158] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HPR
Gene Name
haptoglobin-related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020995.3 175 Missense Mutation GTT,TTT V49F NP_066275.3
Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 175 Intron NP_001135789.1
NM_001142318.1 175 Intron NP_001135790.1
NM_001324354.1 175 Intron NP_001311283.1
NM_001324355.1 175 Intron NP_001311284.1
NM_017853.2 175 Intron NP_060323.1
XM_017023377.1 175 Intron XP_016878866.1

View Full Product Details