Product Details

SNP ID
rs187443721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48878206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAATGGCTGAGTTGTCCGAGCT[A/G]TATGAAGAGAGCAGTGACCTGCAGA
Phenotype
MIM: 605894
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EID1 PubMed Links

Gene Details

Gene
EID1
Gene Name
EP300 interacting inhibitor of differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014335.2 114 Silent Mutation CTA,CTG L10L NP_055150.1
Gene
SHC4
Gene Name
SHC adaptor protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203349.3 114 Intron NP_976224.3
XM_005254375.3 114 Intron XP_005254432.1

View Full Product Details