Product Details

SNP ID
rs187592622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77377634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGAGGGGGAGCCAGACAGTGC[C/T]AAGAACGTGCAGCTGAAACCTGGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAMD15 PubMed Links

Gene Details

Gene
SAMD15
Gene Name
sterile alpha motif domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010860.1 216 Silent Mutation GCC,GCT A72A NP_001010860.1
Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 216 Intron NP_998766.1
XM_017021223.1 216 Intron XP_016876712.1
XM_017021224.1 216 Intron XP_016876713.1
XM_017021225.1 216 Intron XP_016876714.1

View Full Product Details