Product Details

SNP ID
rs187308634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:3311555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGAGCTACGCACAGGTAATCTCC[A/G]GCATCTTTGTCCGTCACTGATTTCA
Phenotype
MIM: 300938
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MXRA5 PubMed Links

Gene Details

Gene
MXRA5
Gene Name
matrix remodeling associated 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015419.3 Intron NP_056234.2
XM_005274485.2 Intron XP_005274542.1

View Full Product Details