Product Details
- SNP ID
-
rs187886723
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:99162670 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTTGCTACAGAGTATGCTGCTT[A/G]TCATCAAATTGATAATCACATTCAC
- Phenotype
-
MIM: 610284
MIM: 607166
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LIPT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2516835] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LIPT1
- Gene Name
- lipoyltransferase 1
- Gene
- MITD1
- Gene Name
- microtubule interacting and trafficking domain containing 1
- Gene
- TSGA10
- Gene Name
- testis specific 10
There are no transcripts associated with this gene.
View Full Product Details