Product Details

SNP ID
rs188324571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:134478659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGCTCGGCAAGCTCCACGGA[C/T]GCTTGCTGGACCGTCTCCTGGATCA
Phenotype
MIM: 610931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFAT PubMed Links

Gene Details

Gene
ZFAT
Gene Name
zinc finger and AT-hook domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029939.3 3900 Silent Mutation NP_001025110.2
NM_001167583.2 3900 Silent Mutation NP_001161055.1
NM_001174157.1 3900 Silent Mutation NP_001167628.1
NM_001174158.1 3900 Silent Mutation NP_001167629.1
NM_001289394.1 3900 Silent Mutation NP_001276323.1
NM_020863.3 3900 Silent Mutation NP_065914.2
XM_011517203.1 3900 Silent Mutation XP_011515505.1
XM_011517204.2 3900 Silent Mutation XP_011515506.1
XM_011517206.1 3900 Missense Mutation XP_011515508.1
XM_017013716.1 3900 Intron XP_016869205.1

View Full Product Details