Product Details

SNP ID
rs188291383
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:47334023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCAGACCCGCCGGCCGATTCT[C/T]CTCTGCTCTCCACGTCTCATGAAGC
Phenotype
MIM: 191710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMPK1 PubMed Links

Gene Details

Gene
CMPK1
Gene Name
cytidine/uridine monophosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136140.1 227 Silent Mutation CTC,CTT L26L NP_001129612.1
NM_016308.2 227 Silent Mutation CTC,CTT L26L NP_057392.1

View Full Product Details