Product Details

SNP ID
rs187988360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99397987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTGCATTTTCATGTAACGCTCT[C/T]TTGCCTTCTGCTTCTCAATCTCTTC
Phenotype
MIM: 604223 MIM: 607075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
There are no transcripts associated with this gene.

Gene
PDAP1
Gene Name
PDGFA associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014891.6 496 Missense Mutation AAA,AGA K121R NP_055706.1

View Full Product Details