Product Details

SNP ID
rs187991947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73555581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCATCCGTGTGATTGATAAC[A/G]TTTTCTATTTAAAAGAAAAGCAATT
Phenotype
MIM: 605787
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 1036 Silent Mutation AAC,AAT N173N NP_075526.1
XM_005248560.2 1036 Silent Mutation AAC,AAT N105N XP_005248617.1
XM_017009678.1 1036 Silent Mutation AAC,AAT N12N XP_016865167.1

View Full Product Details