Product Details
- SNP ID
-
rs188117420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:41466010 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAACTTTGTGGCTCCTCTTTTGAG[G/T]CTCCCTTTTTCCATGTTTTCCCTCC
- Phenotype
-
MIM: 300278
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NYX
PubMed Links
Gene Details
- Gene
- NYX
- Gene Name
- nyctalopin
View Full Product Details