Product Details

SNP ID
rs188451659
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:162378949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGCCCAGGCAGGAGGTGACAC[G/T]CTCAAAGTGACTGGTCACTACGTAA
Phenotype
MIM: 605551
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf111 PubMed Links

Gene Details

Gene
C1orf111
Gene Name
chromosome 1 open reading frame 111
There are no transcripts associated with this gene.

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085375.1 44 Intron NP_001078844.1
NM_001135240.1 44 Silent Mutation ACG,ACT T12T NP_001128712.1
Gene
NOS1AP
Gene Name
nitric oxide synthase 1 adaptor protein
There are no transcripts associated with this gene.

View Full Product Details