Product Details
- SNP ID
-
rs188731444
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:73062512 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACACCAGGCATTACTGAGTGTTG[G/T]TGTGAGCAAGAGGTCTAATACTGTG
- Phenotype
-
MIM: 613250
MIM: 613822
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EBLN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200718150] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EBLN2
- Gene Name
- endogenous Bornavirus-like nucleoprotein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018029.4 |
854 |
Missense Mutation |
GGT,GTT |
G144V |
NP_060499.3 |
- Gene
- PPP4R2
- Gene Name
- protein phosphatase 4 regulatory subunit 2
View Full Product Details