Product Details

SNP ID
rs188383913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:154122063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGTGGCGGAGGCGGTCGATTC[C/T]TCAGCCCCACCAGGTTCTGGGCCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF26 PubMed Links

Gene Details

Gene
ARHGEF26
Gene Name
Rho guanine nucleotide exchange factor 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251962.1 282 Missense Mutation CCT,CTT P24L NP_001238891.1
NM_001251963.1 282 Missense Mutation CCT,CTT P24L NP_001238892.1
NM_015595.3 282 Missense Mutation CCT,CTT P24L NP_056410.3
XM_011512672.1 282 Missense Mutation CCT,CTT P24L XP_011510974.1
XM_017006160.1 282 Intron XP_016861649.1
Gene
ARHGEF26-AS1
Gene Name
ARHGEF26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details