Product Details

SNP ID
rs189382441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18081273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCCACTGGGACGTCTCTTTTCT[C/T]TGAGTTTGAAGTTTATCAGGATGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERGL PubMed Links

Gene Details

Gene
RERGL
Gene Name
RERG like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286201.1 743 Missense Mutation AAA,AGA K178R NP_001273130.1
NM_024730.3 743 Missense Mutation AAA,AGA K179R NP_079006.1

View Full Product Details