Product Details

SNP ID
rs189306575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144399799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCAGGGGCTGCAGGATTGTCC[G/T]TGCTTTCTTTCCCATTCAGTTTCTC
Phenotype
MIM: 610934
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOBOX PubMed Links

Gene Details

Gene
NOBOX
Gene Name
NOBOX oogenesis homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080413.3 1227 Missense Mutation AAG,ACG K371T NP_001073882.3
XM_017011742.1 1227 Missense Mutation AAG,ACG K339T XP_016867231.1

View Full Product Details