Product Details

SNP ID
rs189043504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12675846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGACCGCAGCCGCTCAGTCCT[C/T]GTTCTTCTCATGCATGAAGGCATCC
Phenotype
MIM: 600944 MIM: 609458 MIM: 616850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHPS PubMed Links

Gene Details

Gene
DHPS
Gene Name
deoxyhypusine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206974.1 1234 Missense Mutation AAG,GAG K326E NP_001193903.1
NM_001930.3 1234 Missense Mutation AAG,GAG K368E NP_001921.1
NM_013406.2 1234 Missense Mutation AAG,GAG K321E NP_037538.1
XM_011527770.1 1234 Missense Mutation AAG,GAG K368E XP_011526072.1
XM_011527771.2 1234 Missense Mutation AAG,GAG K185E XP_011526073.1
Gene
MAN2B1
Gene Name
mannosidase alpha class 2B member 1
There are no transcripts associated with this gene.

Gene
WDR83
Gene Name
WD repeat domain 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099737.2 1234 Intron NP_001093207.1
NM_032332.3 1234 Intron NP_115708.1
Gene
WDR83OS
Gene Name
WD repeat domain 83 opposite strand
There are no transcripts associated with this gene.

View Full Product Details