Product Details

SNP ID
rs189122791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123457495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATCTGACCATGACCAGATCACT[A/G]AGATTGTCGCAAGCACTGAGGACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282855.1 2668 Silent Mutation CTC,CTT L693L NP_001269784.1
NM_001321293.1 2668 Silent Mutation CTC,CTT L693L NP_001308222.1
NM_178827.4 2668 Silent Mutation CTC,CTT L693L NP_849149.3
XM_005250161.3 2668 Silent Mutation CTC,CTT L693L XP_005250218.1
XM_005250162.4 2668 Silent Mutation CTC,CTT L693L XP_005250219.1
XM_011515833.2 2668 Silent Mutation CTC,CTT L693L XP_011514135.1
XM_011515834.2 2668 Silent Mutation CTC,CTT L693L XP_011514136.1
XM_017011771.1 2668 Silent Mutation CTC,CTT L693L XP_016867260.1
XM_017011772.1 2668 Silent Mutation CTC,CTT L414L XP_016867261.1

View Full Product Details