Product Details

SNP ID
rs189159887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:40393499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCACTTTGGTGCTTCTTCAAA[G/T]GTAAGAAACTAATTTAAAGAATTCT
Phenotype
MIM: 612170
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC19 PubMed Links

Gene Details

Gene
MUC19
Gene Name
mucin 19, oligomeric
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173600.2 Intron NP_775871.2

View Full Product Details