Product Details

SNP ID
rs188934797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135665891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTACTCCAAGCATTACAGGAGCC[A/C]GCCCAGGCCTCCCATGCGTGAGCTG
Phenotype
MIM: 612903
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCN9 PubMed Links

Gene Details

Gene
LCN9
Gene Name
lipocalin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001676.1 506 Missense Mutation CAG,CCG Q169P NP_001001676.1
XM_017014712.1 506 Missense Mutation CAG,CCG Q169P XP_016870201.1

View Full Product Details