Product Details
- SNP ID
-
rs189771305
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:99612595 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTAATGTTGCCACACACCCGGC[C/T]GCACCTGCAAACAAGAAAACAACTG
- Phenotype
-
MIM: 609767
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A28
PubMed Links
Gene Details
- Gene
- SLC25A28
- Gene Name
- solute carrier family 25 member 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031212.3 |
540 |
Silent Mutation |
GCA,GCG |
A175A |
NP_112489.3 |
XM_005270208.1 |
540 |
Silent Mutation |
GCA,GCG |
A76A |
XP_005270265.1 |
XM_005270211.4 |
540 |
Silent Mutation |
GCA,GCG |
A76A |
XP_005270268.1 |
XM_006718005.3 |
540 |
Silent Mutation |
GCA,GCG |
A76A |
XP_006718068.1 |
XM_006718006.3 |
540 |
Silent Mutation |
GCA,GCG |
A175A |
XP_006718069.1 |
XM_011540239.2 |
540 |
Silent Mutation |
GCA,GCG |
A85A |
XP_011538541.1 |
XM_011540240.1 |
540 |
Silent Mutation |
GCA,GCG |
A76A |
XP_011538542.1 |
XM_011540244.2 |
540 |
Silent Mutation |
GCA,GCG |
A175A |
XP_011538546.1 |
XM_017016741.1 |
540 |
Silent Mutation |
GCA,GCG |
A76A |
XP_016872230.1 |
XM_017016742.1 |
540 |
Silent Mutation |
GCA,GCG |
A20A |
XP_016872231.1 |
View Full Product Details