Product Details

SNP ID
rs189474091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99340611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTTTCCACGTGCGTCCAGTCA[A/G]TAGCAGCATAGGGTTTATTGAGAGG
Phenotype
MIM: 103730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADH1C PubMed Links

Gene Details

Gene
ADH1C
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000669.4 1088 Silent Mutation CTG,TTG L310L NP_000660.1
XM_011531588.2 1088 Silent Mutation CTG,TTG L276L XP_011529890.1
XM_011531589.1 1088 Silent Mutation CTG,TTG L270L XP_011529891.1

View Full Product Details