Product Details

SNP ID
rs190109995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158641492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAATTGGGACTGAGGCAGAAGGA[C/T]AGATTGAGTCCAGGAGTCCAGGAGT
Phenotype
MIM: 608532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCAPG2 PubMed Links

Gene Details

Gene
NCAPG2
Gene Name
non-SMC condensin II complex subunit G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281932.1 3447 Intron NP_001268861.1
NM_001281933.1 3447 Missense Mutation TAT,TGT Y1148C NP_001268862.1
NM_017760.6 3447 Intron NP_060230.5
XM_005249547.2 3447 Intron XP_005249604.1
XM_011516356.2 3447 Missense Mutation TAT,TGT Y1148C XP_011514658.1
XM_011516357.2 3447 Missense Mutation TAT,TGT Y1148C XP_011514659.1
XM_011516358.2 3447 Intron XP_011514660.1
XM_011516362.1 3447 Intron XP_011514664.1
XM_017012374.1 3447 Missense Mutation TAT,TGT Y1101C XP_016867863.1
XM_017012375.1 3447 Missense Mutation TAT,TGT Y1090C XP_016867864.1
XM_017012376.1 3447 Intron XP_016867865.1
XM_017012377.1 3447 Missense Mutation TAT,TGT Y739C XP_016867866.1
XM_017012378.1 3447 Intron XP_016867867.1

View Full Product Details