Product Details

SNP ID
rs190084506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:16475167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATCTTCATCATTTGTTCCCAT[A/G]CCATTTTCTAGAGATGGAAAATCTG
Phenotype
MIM: 613114 MIM: 608694
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM134B PubMed Links

Gene Details

Gene
FAM134B
Gene Name
family with sequence similarity 134 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034850.2 1214 Silent Mutation GGC,GGT G356G NP_001030022.1
NM_019000.4 1214 Silent Mutation GGC,GGT G215G NP_061873.2
XM_011514053.2 1214 Silent Mutation GGC,GGT G396G XP_011512355.1
XM_011514054.2 1214 Silent Mutation GGC,GGT G255G XP_011512356.1
XM_011514055.2 1214 Silent Mutation GGC,GGT G223G XP_011512357.1
Gene
ZNF622
Gene Name
zinc finger protein 622
There are no transcripts associated with this gene.

View Full Product Details