Product Details

SNP ID
rs190422699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100063974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTCTTCAAGAGGCAGGCTCCA[A/T]GAAAGGTAGAGAATCAGTTCCTACT
Phenotype
MIM: 194510 MIM: 601261
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF3 PubMed Links

Gene Details

Gene
ZNF3
Gene Name
zinc finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278284.1 923 Intron NP_001265213.1
NM_001278287.1 923 Intron NP_001265216.1
NM_001278290.1 923 Intron NP_001265219.1
NM_001278291.1 923 Intron NP_001265220.1
NM_001278292.1 923 Intron NP_001265221.1
NM_001318135.1 923 Intron NP_001305064.1
NM_001318136.1 923 Intron NP_001305065.1
NM_001318137.1 923 UTR 3 NP_001305066.1
NM_017715.3 923 UTR 3 NP_060185.2
NM_032924.4 923 Intron NP_116313.3
XM_006716111.1 923 Intron XP_006716174.1
XM_017012582.1 923 Intron XP_016868071.1
XM_017012583.1 923 Intron XP_016868072.1
Gene
ZSCAN21
Gene Name
zinc finger and SCAN domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145914.2 923 Missense Mutation AAG,ATG K260M NP_666019.1
XM_005250568.4 923 Missense Mutation AAG,ATG K260M XP_005250625.1
XM_006716113.3 923 Missense Mutation AAG,ATG K260M XP_006716176.1
XM_017012584.1 923 Missense Mutation AAG,ATG K260M XP_016868073.1
XM_017012585.1 923 Missense Mutation AAG,ATG K260M XP_016868074.1

View Full Product Details