Product Details
- SNP ID
-
rs190419930
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:134987959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTCAGAAGATTCAGAAAAGGAAG[A/G]CTATTCAGACAGAACAATCAGTGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AGBL3
PubMed Links
Gene Details
- Gene
- AGBL3
- Gene Name
- ATP/GTP binding protein like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178563.3 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
NP_848658.3 |
XM_011516122.2 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_011514424.1 |
XM_017012136.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867625.1 |
XM_017012137.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867626.1 |
XM_017012138.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867627.1 |
XM_017012139.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867628.1 |
XM_017012140.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867629.1 |
XM_017012141.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867630.1 |
XM_017012142.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867631.1 |
XM_017012143.1 |
1601 |
Missense Mutation |
GAC,GGC |
D9G |
XP_016867632.1 |
View Full Product Details