Product Details

SNP ID
rs190254963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49946158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACCAATTGGTTTTTCAAAATC[A/G]TGACACTCATGTCGACATTTGCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB133 PubMed Links

Gene Details

Gene
DEFB133
Gene Name
defensin beta 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166478.1 129 Silent Mutation CAC,CAT H43H NP_001159950.1
XM_017010860.1 129 Silent Mutation CAC,CAT H43H XP_016866349.1

View Full Product Details