Product Details

SNP ID
rs190890755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104491193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACTTGCACATGCTCCTACTGTC[A/G]CTTCTACTGCCGTTGTGGGAGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP12 PubMed Links

Gene Details

Gene
LRP12
Gene Name
LDL receptor related protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135703.2 2187 Missense Mutation GCG,GTG A668V NP_001129175.1
NM_013437.4 2187 Missense Mutation GCG,GTG A687V NP_038465.1
Gene
MIR548A3
Gene Name
microRNA 548a-3
There are no transcripts associated with this gene.

View Full Product Details