Product Details

SNP ID
rs191037362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85109674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATCTCCAAGATCGAAGCCATT[G/T]ATCATATTTGGAATTTACAACATCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRCC1 PubMed Links

Gene Details

Gene
LRRCC1
Gene Name
leucine rich repeat and coiled-coil centrosomal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033402.4 80 Missense Mutation GAT,TAT D62Y NP_208325.3
XM_005251318.3 80 Intron XP_005251375.1
XM_005251319.4 80 UTR 5 XP_005251376.1
XM_005251320.3 80 Intron XP_005251377.1
XM_005251322.3 80 Intron XP_005251379.1
XM_017013920.1 80 Missense Mutation GAT,TAT D62Y XP_016869409.1
XM_017013921.1 80 Missense Mutation GAT,TAT D62Y XP_016869410.1
XM_017013922.1 80 Missense Mutation GAT,TAT D62Y XP_016869411.1
XM_017013923.1 80 Intron XP_016869412.1
XM_017013924.1 80 Missense Mutation GAT,TAT D62Y XP_016869413.1
XM_017013925.1 80 UTR 5 XP_016869414.1
XM_017013926.1 80 Intron XP_016869415.1
XM_017013927.1 80 Intron XP_016869416.1
XM_017013928.1 80 Intron XP_016869417.1

View Full Product Details