Product Details

SNP ID
rs190738237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55452868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTGATTCTGTCCTACTTGTAC[A/G]TAGTCAGAACAATTCTGAAGTTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6C2 PubMed Links

Gene Details

Gene
OR6C2
Gene Name
olfactory receptor family 6 subfamily C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054105.1 655 Missense Mutation ATA,GTA I219V NP_473446.1

View Full Product Details